siRNA Details
Gene Symbol: Olfr199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207550.2 | NP_997433.2 | 1 | 295 | ||
GenBank mRNA | BC128028.1 | - | 1 | 299 | 40136725 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207550.2 | NP_997433.2 | 1 | 295 | ||
GenBank mRNA | BC128028.1 | - | 1 | 299 | 40136725 |