siRNA Details
| Gene Symbol: RCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271837.1 | NP_001258766.1 | 3 | 542 | ||
| NM_002902.2 | NP_002893.1 | 3 | 542 | |||
| GenBank mRNA | AK313833.1 | - | 3 | 413 | ||
| AL110102.1 | - | 2 | 119 | |||
| BC004892.2 | - | 3 | 382 | 3505241 | ||
| BP275421.1 | - | 3 | 408 | |||
| BT009790.1 | - | 3 | 321 | |||
| X78669.1 | - | 3 | 387 | |||

RCN2
Genomic Map