siRNA Details
| Gene Symbol: Olfr251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207553.2 | NP_997436.2 | 1 | 296 | ||
| Gene Symbol: Olfr900 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146874.1 | NP_667085.1 | 1 | 296 | ||
| GenBank mRNA | BC106986.2 | - | 1 | 296 | 40043276 | |

Olfr251
Genomic Map