siRNA Details
| Gene Symbol: Olfr372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207555.2 | NP_997438.1 | 1 | 923 | ||
| GenBank mRNA | BC150671.1 | - | 1 | 923 | 9087582 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr372| Gene Symbol: Olfr372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207555.2 | NP_997438.1 | 1 | 923 | ||
| GenBank mRNA | BC150671.1 | - | 1 | 923 | 9087582 | |