siRNA Details
| Gene Symbol: Olfr403 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207622.1 | NP_997505.1 | 1 | 229 | ||
| GenBank mRNA | AF102517.1 | - | 1 | 40 | ||
| BC104289.1 | - | 1 | 229 | 40042769 | ||
| BC104290.1 | - | 1 | 229 | 40042770 | ||
| Gene Symbol: Olfr43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146711.2 | NP_666922.2 | 1 | 304 | ||
| GenBank mRNA | BC125464.1 | - | 1 | 304 | 40129979 | |
| BC125468.1 | - | 1 | 304 | 40129983 | ||

Olfr403
Genomic Map