siRNA Details
| Gene Symbol: Olfr681 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207557.2 | NP_997440.2 | 1 | 335 | ||
| GenBank mRNA | BC104298.1 | - | 1 | 335 | 40043142 | |
| BC104299.1 | - | 1 | 335 | 40043148 | ||
| Gene Symbol: Olfr679 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147044.1 | NP_667255.1 | 1 | 335 | ||
| GenBank mRNA | BC127993.1 | - | 1 | 339 | 40135720 | |

Olfr681
Genomic Map