siRNA Details
| Gene Symbol: RCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271837.1 | NP_001258766.1 | 5, 6 | 843 | ||
| NM_002902.2 | NP_002893.1 | 4, 5 | 789 | |||
| GenBank mRNA | AK313833.1 | - | 4, 5 | 660 | ||
| AL110102.1 | - | 3, 4 | 366 | |||
| AV711990.1 | - | 3, 4 | 294 | |||
| BC004892.2 | - | 4, 5 | 629 | 3505241 | ||
| BT009790.1 | - | 4, 5 | 568 | |||
| X78669.1 | - | 4, 5 | 634 | |||

RCN2
Genomic Map