siRNA Details
| Gene Symbol: Olfr924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207560.1 | NP_997443.1 | 1 | 411 | ||
| Gene Symbol: Olfr926 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146815.1 | NP_667026.1 | 1 | 411 | ||
| GenBank mRNA | BC120558.1 | - | 1 | 463 | 40129481 | |
| BC125490.1 | - | 1 | 463 | 40130005 | ||

Olfr924
Genomic Map