siRNA Details
| Gene Symbol: Olfr1113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207565.1 | NP_997448.1 | 1 | 623 | ||
| Gene Symbol: Olfr1115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146297.2 | NP_666409.2 | 1 | 722 | ||
| GenBank mRNA | BC141024.1 | - | 1 | 722 | 9055636 | |

Olfr1113
Genomic Map