siRNA Details
| Gene Symbol: Olfr1383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207574.1 | NP_997457.1 | 1 | 148 | ||
| GenBank mRNA | BC127965.1 | - | 1 | 152 | 40134985 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1383| Gene Symbol: Olfr1383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207574.1 | NP_997457.1 | 1 | 148 | ||
| GenBank mRNA | BC127965.1 | - | 1 | 152 | 40134985 | |