siRNA Details
| Gene Symbol: Olfr322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207693.1 | NP_997576.1 | 1 | 756 | ||
| Gene Symbol: Olfr317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011769.2 | NP_001011769.2 | 1 | 816 | ||
| GenBank mRNA | BC147131.1 | - | 1 | 816 | 9056194 | |
| BC147132.1 | - | 1 | 816 | 9056174 | ||

Olfr322
Genomic Map