siRNA Details
Gene Symbol: Olfr322 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207693.1 | NP_997576.1 | 1 | 238 |
Gene Symbol: Olfr317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC147131.1 | - | 1 | 298 | 9056194 | |
BC147132.1 | - | 1 | 298 | 9056174 |
