siRNA Details
| Gene Symbol: Olfr1082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207674.2 | NP_997557.2 | 2 | 877 | ||
| Gene Symbol: Olfr1084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207135.2 | NP_997018.2 | 1 | 841 | ||

Olfr1082
Genomic Map