siRNA Details
| Gene Symbol: Bex4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212457.2 | NP_997622.1 | 3 | 413 | ||
| GenBank mRNA | AK010400.1 | - | 3 | 416 | ||
| AY833558.1 | - | 3 | 413 | |||
| BC090663.1 | - | 3 | 390 | 30677421 | ||
| BC116226.2 | - | 3 | 317 | 40091511 | ||

Bex4
Genomic Map