siRNA Details
| Gene Symbol: Krt83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003668.2 | NP_001003668.2 | 5 | 824 | ||
| GenBank mRNA | BC139321.1 | - | 5 | 824 | 8862343 | |
| BC139322.1 | - | 5 | 824 | 8862344 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt83| Gene Symbol: Krt83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003668.2 | NP_001003668.2 | 5 | 824 | ||
| GenBank mRNA | BC139321.1 | - | 5 | 824 | 8862343 | |
| BC139322.1 | - | 5 | 824 | 8862344 | ||