siRNA Details
| Gene Symbol: Krt40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039666.1 | NP_001034755.1 | 1 | 252 | ||
| GenBank mRNA | BC107359.1 | - | 1 | 252 | ||
| BC107360.2 | - | 1 | 250 | |||
| BC119521.2 | - | 1 | 252 | 40055957 | ||
| BC119522.2 | - | 1 | 252 | 40055959 | ||

Krt40
Genomic Map