siRNA Details
| Gene Symbol: BC051142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001177.2 | NP_001001177.2 | 6 | 697 | ||
| GenBank mRNA | AK077077.1 | - | 6 | 684 | ||
| AK133031.1 | - | 6 | 545 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC051142| Gene Symbol: BC051142 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001177.2 | NP_001001177.2 | 6 | 697 | ||
| GenBank mRNA | AK077077.1 | - | 6 | 684 | ||
| AK133031.1 | - | 6 | 545 | |||