siRNA Details
| Gene Symbol: BC051408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC051408.1 | - | 2 | 241 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC051408| Gene Symbol: BC051408 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC051408.1 | - | 2 | 241 | ||