siRNA Details
| Gene Symbol: Zfp874b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076791.2 | NP_001070259.1 | 4 | 547 | ||
| GenBank mRNA | AK079330.1 | - | 2 | 253 | ||
| AK137581.1 | - | 4 | 525 | |||
| BY228006.1 | - | 2 | 331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp874b| Gene Symbol: Zfp874b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001076791.2 | NP_001070259.1 | 4 | 547 | ||
| GenBank mRNA | AK079330.1 | - | 2 | 253 | ||
| AK137581.1 | - | 4 | 525 | |||
| BY228006.1 | - | 2 | 331 | |||