siRNA Details
| Gene Symbol: Wfdc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012704.1 | NP_001012722.1 | 2 | 250 | ||
| GenBank mRNA | AY542486.1 | - | 2 | 250 | ||
| AY542487.1 | - | 2 | 250 | |||
| BC145778.1 | - | 2 | 159 | 40131081 | ||
| BC145780.1 | - | 2 | 159 | 40131083 | ||

Wfdc13
Genomic Map