siRNA Details
| Gene Symbol: Gm5415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164286.1 | NP_001157758.1 | 1 | 2173 | ||
| GenBank mRNA | AK029494.1 | - | 1 | 2175 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5415| Gene Symbol: Gm5415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164286.1 | NP_001157758.1 | 1 | 2173 | ||
| GenBank mRNA | AK029494.1 | - | 1 | 2175 | ||