siRNA Details
Gene Symbol: Gm5416 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082542.1 | NP_001076011.1 | 2 | 153 | ||
GenBank mRNA | BC115936.1 | - | 2 | 152 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5416 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082542.1 | NP_001076011.1 | 2 | 153 | ||
GenBank mRNA | BC115936.1 | - | 2 | 152 |