siRNA Details
| Gene Symbol: BC024978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243888.1 | NP_001230817.1 | 6 | 1142 | ||
| NM_001290511.1 | NP_001277440.1 | 6 | 1142 | |||
| NM_001290513.1 | NP_001277442.1 | 6 | 1142 | |||
| GenBank mRNA | AK144930.1 | - | 1 | 25 | ||
| AK166513.1 | - | 7 | 989 | |||
| BC024978.1 | - | 5 | 598 | |||

BC024978
Genomic Map