siRNA Details
| Gene Symbol: Itprip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001738.2 | NP_001001738.2 | 2 | 1143 | ||
| GenBank mRNA | AK160686.1 | - | 2 | 1145 | ||
| AK165192.1 | - | 2 | 1122 | |||
| AK171522.1 | - | 2 | 1049 | |||
| AK171539.1 | - | 2 | 1049 | |||
| AK171911.1 | - | 2 | 1124 | |||
| AK220222.1 | - | 2 | 1057 | |||
| BC063749.1 | - | 3 | 1109 | 6813779 | ||

Itprip
Genomic Map