siRNA Details
| Gene Symbol: Zyg11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033634.3 | NP_001028806.2 | 7 | 1559 | ||
| GenBank mRNA | AK145130.1 | - | 7 | 1533 | ||
| AK148335.1 | - | 7 | 1558 | |||
| AK148760.1 | - | 1 | 29 | |||
| AK166031.1 | - | 7 | 1539 | |||
| AK166174.1 | - | 7 | 1539 | |||
| AK166278.1 | - | 7 | 1550 | |||
| BC117977.2 | - | 7 | 1523 | 40099428 | ||
| BC117978.2 | - | 7 | 1523 | 40099429 | ||
| BC139383.1 | - | 7 | 1526 | 8862405 | ||
| BC139384.1 | - | 7 | 1526 | 8862406 | ||

Zyg11b
Genomic Map