siRNA Details
| Gene Symbol: Gnptab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004164.2 | NP_001004164.2 | 14 | 2840 | ||
| GenBank mRNA | AK140867.1 | - | 13 | 2944 | ||
| AK154710.1 | - | 6 | 1739 | |||
| AK173132.1 | - | 14 | 2840 | |||
| BC060638.1 | - | 16 | 2324 | 6849405 | ||
| L36434.1 | - | 2 | 910 | |||

Gnptab
Genomic Map