siRNA Details
| Gene Symbol: Specc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029936.3 | NP_001025107.1 | 4 | 2045 | ||
| NM_001281818.1 | NP_001268747.1 | 6 | 2262 | |||
| GenBank mRNA | AK220465.1 | - | 4 | 1972 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Specc1| Gene Symbol: Specc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029936.3 | NP_001025107.1 | 4 | 2045 | ||
| NM_001281818.1 | NP_001268747.1 | 6 | 2262 | |||
| GenBank mRNA | AK220465.1 | - | 4 | 1972 | ||