siRNA Details
| Gene Symbol: Krtap31-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025244.3 | NP_001020415.1 | 1 | 799 | ||
| GenBank mRNA | AK037252.1 | - | 1 | 802 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap31-2| Gene Symbol: Krtap31-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025244.3 | NP_001020415.1 | 1 | 799 | ||
| GenBank mRNA | AK037252.1 | - | 1 | 802 | ||