siRNA Details
Gene Symbol: Dnaic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034878.2 | NP_001030050.2 | 12 | 2009 | ||
GenBank mRNA | CJ049712.1 | - | 2 | 302 | ||
EF565270.1 | - | 11 | 1625 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnaic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034878.2 | NP_001030050.2 | 12 | 2009 | ||
GenBank mRNA | CJ049712.1 | - | 2 | 302 | ||
EF565270.1 | - | 11 | 1625 |