siRNA Details
| Gene Symbol: Zscan26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013786.2 | NP_001013808.2 | 4 | 854 | ||
| GenBank mRNA | BC068174.1 | - | 2 | 678 | 30547278 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zscan26| Gene Symbol: Zscan26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013786.2 | NP_001013808.2 | 4 | 854 | ||
| GenBank mRNA | BC068174.1 | - | 2 | 678 | 30547278 | |