siRNA Details
| Gene Symbol: Vmn1r209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013787.1 | NP_001013809.1 | 1 | 494 | ||
| Gene Symbol: Vmn1r206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134216.1 | NP_598977.1 | 1 | 494 | ||
| GenBank mRNA | BC132625.1 | - | 1 | 594 | 40130902 | |
| BC138067.1 | - | 1 | 594 | 8861086 | ||

Vmn1r209
Genomic Map