siRNA Details
| Gene Symbol: Prr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030296.4 | NP_001025467.1 | 2 | 523 | ||
| GenBank mRNA | AK133129.1 | - | 2 | 483 | ||
| BC138854.1 | - | 1 | 191 | 8861876 | ||
| BC138856.1 | - | 1 | 191 | 8861878 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr7| Gene Symbol: Prr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030296.4 | NP_001025467.1 | 2 | 523 | ||
| GenBank mRNA | AK133129.1 | - | 2 | 483 | ||
| BC138854.1 | - | 1 | 191 | 8861876 | ||
| BC138856.1 | - | 1 | 191 | 8861878 | ||