siRNA Details
| Gene Symbol: Prr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030296.4 | NP_001025467.1 | 3 | 1315 | ||
| GenBank mRNA | AK133129.1 | - | 3 | 1275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr7| Gene Symbol: Prr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030296.4 | NP_001025467.1 | 3 | 1315 | ||
| GenBank mRNA | AK133129.1 | - | 3 | 1275 | ||