siRNA Details
| Gene Symbol: Gm5458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024706.2 | NP_001019877.2 | 2 | 230 | ||
| GenBank mRNA | BC060995.1 | - | 2 | 230 | 6774503 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5458| Gene Symbol: Gm5458 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024706.2 | NP_001019877.2 | 2 | 230 | ||
| GenBank mRNA | BC060995.1 | - | 2 | 230 | 6774503 | |