siRNA Details
| Gene Symbol: Otulin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013792.2 | NP_001013814.2 | 6 | 787 | ||
| GenBank mRNA | AK134955.1 | - | 6 | 622 | ||
| AK150371.1 | - | 6 | 790 | |||
| BC087945.1 | - | 3 | 388 | 30505408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otulin| Gene Symbol: Otulin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013792.2 | NP_001013814.2 | 6 | 787 | ||
| GenBank mRNA | AK134955.1 | - | 6 | 622 | ||
| AK150371.1 | - | 6 | 790 | |||
| BC087945.1 | - | 3 | 388 | 30505408 | ||