siRNA Details
| Gene Symbol: A930007A09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044312.1 | - | 1 | 259 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930007A09Rik| Gene Symbol: A930007A09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044312.1 | - | 1 | 259 | ||