siRNA Details
| Gene Symbol: Gm5483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082547.1 | NP_001076016.1 | 1, 2 | 107 | ||
| GenBank mRNA | AK089257.1 | - | 1, 2 | 107 | ||
| BB843120.1 | - | 1, 2 | 107 | |||
| BC147081.1 | - | 1, 2 | 88 | 9056150 | ||
| BC147082.1 | - | 1, 2 | 88 | 9056148 | ||

Gm5483
Genomic Map