siRNA Details
| Gene Symbol: Plcxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134480.1 | NP_001127952.1 | 2 | 1045 | ||
| GenBank mRNA | BC151059.1 | - | 2 | 647 | 9087972 | |
| BC151063.1 | - | 2 | 647 | 9087976 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plcxd2| Gene Symbol: Plcxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134480.1 | NP_001127952.1 | 2 | 1045 | ||
| GenBank mRNA | BC151059.1 | - | 2 | 647 | 9087972 | |
| BC151063.1 | - | 2 | 647 | 9087976 | ||