siRNA Details
| Gene Symbol: Mettl21c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013799.1 | NP_001013821.1 | 5 | 507 | ||
| GenBank mRNA | AK041301.1 | - | 5 | 507 | ||
| BC147328.1 | - | 5 | 445 | 9056394 | ||
| BC147329.1 | - | 5 | 445 | 9056414 | ||

Mettl21c
Genomic Map