siRNA Details
| Gene Symbol: Teddm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008426.3 | NP_001008426.1 | 1 | 196 | ||
| GenBank mRNA | AK033834.1 | - | 1 | 197 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Teddm1b| Gene Symbol: Teddm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008426.3 | NP_001008426.1 | 1 | 196 | ||
| GenBank mRNA | AK033834.1 | - | 1 | 197 | ||