siRNA Details
| Gene Symbol: Olfr1197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005225.1 | NP_001005225.1 | 1 | 758 | ||
| Gene Symbol: Olfr1200 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005227.2 | NP_001005227.2 | 1 | 758 | ||

Olfr1197
Genomic Map