siRNA Details
| Gene Symbol: AA467197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004174.1 | NP_001004174.1 | 3 | 180 | ||
| GenBank mRNA | AK141297.1 | - | 2 | 283 | ||
| AK150561.1 | - | 3 | 170 | |||
| BC049633.1 | - | 3 | 180 | 6774154 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AA467197| Gene Symbol: AA467197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004174.1 | NP_001004174.1 | 3 | 180 | ||
| GenBank mRNA | AK141297.1 | - | 2 | 283 | ||
| AK150561.1 | - | 3 | 170 | |||
| BC049633.1 | - | 3 | 180 | 6774154 | ||