siRNA Details
| Gene Symbol: AA467197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004174.1 | NP_001004174.1 | 4, 5 | 271 | ||
| GenBank mRNA | AK141297.1 | - | 3, 4 | 374 | ||
| AK150561.1 | - | 4, 5 | 261 | |||
| BC049633.1 | - | 4, 5 | 271 | 6774154 | ||

AA467197
Genomic Map