siRNA Details
| Gene Symbol: Pgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008828.3 | NP_032854.2 | 6 | 712 | ||
| GenBank mRNA | AK133877.1 | - | 6 | 634 | ||
| AK164440.1 | - | 6 | 634 | |||
| AK167441.1 | - | 6 | 606 | |||
| AK167459.1 | - | 6 | 606 | |||
| AK167710.1 | - | 6 | 605 | |||
| AK207010.1 | - | 2 | 131 | |||
| BC023819.1 | - | 6 | 618 | |||
| BC032167.1 | - | 6 | 618 | |||
| BC083355.1 | - | 6 | 604 | 6828087 | ||
| BC108372.1 | - | 6 | 616 | 5060103 | ||
| M15668.1 | - | 6 | 583 | |||

Pgk1
Genomic Map