siRNA Details
| Gene Symbol: Gm5538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101531.1 | NP_001095001.1 | 2, 3 | 400 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5538| Gene Symbol: Gm5538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101531.1 | NP_001095001.1 | 2, 3 | 400 | ||