siRNA Details
| Gene Symbol: Kprp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028629.1 | NP_082905.1 | 2 | 1799 | ||
| GenBank mRNA | AK003253.1 | - | 1 | 89 | ||
| BC141372.1 | - | 2 | 1782 | 9055985 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kprp| Gene Symbol: Kprp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028629.1 | NP_082905.1 | 2 | 1799 | ||
| GenBank mRNA | AK003253.1 | - | 1 | 89 | ||
| BC141372.1 | - | 2 | 1782 | 9055985 | ||