siRNA Details
| Gene Symbol: Gm5547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA146509.1 | - | 2 | 228 | ||
| AK138691.1 | - | 3 | 255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5547| Gene Symbol: Gm5547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA146509.1 | - | 2 | 228 | ||
| AK138691.1 | - | 3 | 255 | |||