siRNA Details
| Gene Symbol: I830077J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033780.3 | NP_001028952.1 | 5, 6 | 364 | ||
| GenBank mRNA | AK152510.1 | - | 5, 6 | 367 | ||
| BC147821.1 | - | 5, 6 | 304 | 9087538 | ||
| BC147828.1 | - | 5, 6 | 304 | 9087545 | ||

I830077J02Rik
Genomic Map