siRNA Details
| Gene Symbol: Ankrd13c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013806.1 | NP_001013828.1 | 13 | 1638 | ||
| GenBank mRNA | AK135906.1 | - | 14 | 1374 | ||
| AK157542.1 | - | 1820 | ||||
| BC065088.1 | - | 12 | 3832 | |||
| BC138991.1 | - | 13 | 1646 | 8862013 | ||
| BC138992.1 | - | 14 | 1625 | 8862014 | ||
| CF736962.1 | - | 6 | 634 | |||

Ankrd13c
Genomic Map