siRNA Details
| Gene Symbol: Spag8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007463.1 | NP_001007464.1 | 2 | 161 | ||
| NM_001290462.1 | NP_001277391.1 | 2 | 161 | |||
| GenBank mRNA | AK132968.1 | - | 2 | 162 | ||
| AY769082.1 | - | 2 | 161 | |||
| BC150868.1 | - | 2 | 203 | 9087779 | ||

Spag8
Genomic Map